missing translation for 'onlineSavingsMsg'
Learn More

ADAM15 Antibody (001), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18328810 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328810 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328810 Supplier Novus Biologicals Supplier No. NBP290765AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ADAM15 Monoclonal specifically detects ADAM15 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM15
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ADAM15 (O88839-1; Met1-Thr697).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8751
Test Specificity Has cross-reactivity in ELISA with: Mouse ADAM9 No cross-reactivity in ELISA with: Human ADAM15
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.