missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM12 Rabbit anti-Human, Clone: 737, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89702-50UL
This item is not returnable.
View return policy
Description
ADAM12 Monoclonal specifically detects ADAM12 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAM12 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8038 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 737 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate | |
| a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.- | |
| Recombinant Human ADAM12 protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction