missing translation for 'onlineSavingsMsg'
Learn More

ADAM12 Antibody (1), DyLight 488 , Novus Biologicals™

Artikelnummer. 18431888 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
Pakningsstørrelse:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18431888 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18431888 Leverandør Novus Biologicals Leverandørnr. NBP289701G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Monoclonal Antibody

ADAM12 Monoclonal antibody specifically detects ADAM12 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ADAM12
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias a disintegrin and metalloproteinase domain 12 (meltrin alpha), ADAM 12, ADAM metallopeptidase domain 12, disintegrin and metalloproteinase domain-containing protein 12, EC 3.4.24, EC 3.4.24.58, MCMP, MCMPMltna, Meltrin-alpha, MLTNA, MLTNEC 3.4.24.-
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ADAM12 extracellular domain (Met 1-Ser 513).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8038
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.