missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550593
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ADAM11 protein.
Probable ligand for integrin in the brain. This is a non catalytic metalloprotease-like protein.
Specifications
| ADAM11 | |
| Polyclonal | |
| Unconjugated | |
| ADAM11 | |
| a disintegrin and metallopeptidase domain 11; a disintegrin and metalloprotease domain (ADAM) 11; a disintegrin and metalloprotease domain 11; a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAM metallopeptidase domain 11; Adam11; ADAMs; AW060611; disintegrin and metalloproteinase domain-containing protein 11; Mdc; metalloendopeptidases; metalloproteinase-like, disintegrin-like, and cysteine-rich protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11488, 4185 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O75078, Q9R1V4 | |
| ADAM11 | |
| Synthetic peptide corresponding to a region derived from internal residues of human ADAM metallopeptidase domain 11. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction