missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581301
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat ADAM10.
Proinflammatory cytokine tumor necrosis factor-a (TNF-a) contributes to a variety of inflammatory responses and programmed cell death. Notch receptor and its ligand participate in cell fate decisions during vertebrate development and are associated with several human disorders, including a T-cell lymphoma. TNF-a, notch and its ligand delta are all membrane-bond molecules, which are cleaved by proteases to release mature proteins or functional receptor. ADAM10, a metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease), was recently identified to cleave TNF-a, notch and its ligand delta (1-3). The genes encoding human, mouse, and bovine ADAM10 were recently cloned and designated ADAM 10, kuzbanian (KUZ), and MADM, respectively. ADAM10 mRNA is expressed in a variety of human and bovine tissues.
Specifications
| ADAM10 | |
| Polyclonal | |
| Unconjugated | |
| Adam10 | |
| 1700031C13Rik; a disintegrin and metallopeptidase domain 10; a disintegrin and metalloprotease domain (ADAM) 10; a disintegrin and metalloprotease domain 10; a disintegrin and metalloproteinase; AD10; AD18; ADAM; ADAM 10; ADAM metallopeptidase domain 10; ADAM10; ADAMs; CD156c; CDw156; Disintegrin and metalloproteinase domain-containing protein 10; HsT18717; KUZ; kuzbanian; kuzbanian protein homolog; MADM; Mammalian disintegrin-metalloprotease; metalloendopeptidases; metalloproteinase; myelin-associated metalloproteinase; RAK | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 29650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q10743 | |
| Adam10 | |
| A synthetic peptide corresponding to the C-terminus of the Rat ADAM10. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction