missing translation for 'onlineSavingsMsg'
Learn More

ADAM10 Antibody (RM0146-7H12), DyLight 594, Novus Biologicals™

Product Code. 18414187 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414187 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414187 Supplier Novus Biologicals Supplier No. NBP212015DL594

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

ADAM10 Monoclonal antibody specifically detects ADAM10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM10
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0146-7H12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease
Host Species Rat
Immunogen Mouse recombinant ADAM10
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 102
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.