Learn More
ADAM10 Antibody (CL13808) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24577C
Product Code. 30305848
  • £558.00 / 0.10mL
Description
ADAM10 Monoclonal antibody specifically detects ADAM10 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ADAM10
Monoclonal
DyLight 650
a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease
This antibody was developed using a synthetic peptide derived from O14672, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Signal Transduction
102
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13808
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified