missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADA2a Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92547-0.1ml
This item is not returnable.
View return policy
Description
ADA2a Polyclonal antibody specifically detects ADA2a in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ADA2a | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ADA2AFLJ12705, ADA2-like protein, hADA2, TADA2LADA2, transcriptional adapter 2-alpha, Transcriptional adapter 2-like, transcriptional adaptor 2 (ADA2 homolog, yeast)-like, transcriptional adaptor 2 alpha, transcriptional adaptor 2A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-443 of human TADA2A (NP_001159577.2). MDRLGSFSNDPSDKPPCRGCSSYLMEPYIKCAECGPPPFFLCLQCFTRGFEYKKHQSDHTYEIMTSDFPVLDPSWTAQEEMALLEAVMDCGFGNWQDVANQMCTKTKEECEKHYMKHFINNPLFASTLLNLKQAEEAKTADTAIPFHSTDDPPRPTFDSLLSRDMAGYMPARADFIEEFDNYAEWDLRDIDFVEDDSDILHALKMAVVDIYHSRLKERQRRKKIIRDHGLINLRKFQLMERRYPKEVQDLYETMRRFARIVGPVEHDKFIESHALEFELRREIKRLQEYRTAGITNFCSARTYDHLKKTREEERLKRTMLSEVLQYIQDSSACQQWLRRQADIDSGLSPSIPMASNSGRRSAPPLNLTGLPGTEKLNEKEKELCQMVRLVPGAYLEYKSALLNECNKQGGLRLAQARALIKIDVNKTRKIYDFLIREGYITKG | |
| 0.1 mL | |
| Cell Cycle and Replication, mTOR Pathway | |
| 6871 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction