missing translation for 'onlineSavingsMsg'
Learn More

ADA2a Antibody (4A8-1A7), Novus Biologicals™

Product Code. 18400750 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400750 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400750 Supplier Novus Biologicals Supplier No. H00006871M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADA2a Monoclonal antibody specifically detects ADA2a in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADA2a
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4A8-1A7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01172
Gene Alias ADA2AFLJ12705, ADA2-like protein, hADA2, TADA2LADA2, transcriptional adapter 2-alpha, Transcriptional adapter 2-like, transcriptional adaptor 2 (ADA2 homolog, yeast)-like, transcriptional adaptor 2 alpha, transcriptional adaptor 2A
Host Species Mouse
Immunogen TADA2L (AAH01172, 1 a.a. ~ 305 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDRLGSFSNDPSDKPPCRGCSSYLMEPYIKCAECGPPPFFLCLQCFTRGFEYKKHRSDHTYEIMTSDFPVLDPSWTAQEEMALLEAVMDCGFGNWQDVANQMCTKTKEECEKHYMKYFINNPLFASTLLNLKQAEEAKTADTAIPFHSTDDPPRPTFDSLLSRDMAGYMPARADFIEEFDNYAEWDLRDIDFVEDDSDILHALKMAVVDIYHSRLKERQRRKKIIRDHGLINLRKFQLMERRYPKEVQDLYETMRRFARIVGPVEHDKFIESHACRWFLSLEQYLCVYIYINRRDNGVFYVKFYK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6871
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.