missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ADA, 99%, for biochemistry

Product Code. 10145140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10145140 25 g Glass bottle 25g
10321044 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10145140 Supplier Thermo Scientific Chemicals Supplier No. 228010250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26239-55-4
Molecular Formula C6H10N2O5
Molecular Weight (g/mol) 190.155
MDL Number MFCD00008031
InChI Key QZTKDVCDBIDYMD-UHFFFAOYSA-N
Synonym N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid
PubChem CID 117765
ChEBI CHEBI:43960
IUPAC Name 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)O

Specifications

Absorbance (9% soln. in 1 M NaOH), (9% soln. in 1 M NaOH) at 0nm, 0.08 max. at 300nm
Absorbance Conditions (9% soln. in 1M NaOH)
Melting Point 219.0°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (110°C, 5 hrs)
Linear Formula H2NCOCH2N(CH2CO2H)2
Packaging Glass bottle
Merck Index 15, 135
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene, chloroform
Formula Weight 190.16
Percent Purity 99%
Grade Biochemical
Chemical Name or Material ADA, For biochemistry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.