missing translation for 'onlineSavingsMsg'
Learn More

ADA, 98+%, for biochemistry, Thermo Scientific™

Product Code. 10751745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
25g; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10751745 25g; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10751745 Supplier Thermo Scientific Acros Supplier No. 327680250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
TRUSTED_SUSTAINABILITY

Specifications

CAS 26239-55-4
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Molecular Formula C6H10N2O5
Linear Formula H2NCOCH2N(CH2CO2H)2
MDL Number MFCD00008031
Quantity 25g
Merck Index 15, 135
Synonym n-2-acetamido iminodiacetic acid, 2,2'-2-amino-2-oxoethyl azanediyl diacetic acid, carbamoylmethylaminodiacetic acid, ada, 2,2'-2-amino-2-oxoethyl imino diacetic acid, n-carbamoylmethyl iminodiacetic acid, glycine, n-2-amino-2-oxoethyl-n-carboxymethyl, n-carbamoylmethyliminodi acetic acid, unii-5c4r3o704e
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene, chloroform
InChI Key QZTKDVCDBIDYMD-UHFFFAOYSA-N
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)O
IUPAC Name 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol) 190.16
PubChem CID 117765
ChEBI CHEBI:43960
Formula Weight 190.16
Percent Purity 98+%
Grade Biochemistry
Chemical Name or Material ADA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.