missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ADA, 98+%

Product Code. 11412328
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412328 25 g 25g
11422328 100 g 100g
11432328 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412328 Supplier Thermo Scientific Chemicals Supplier No. A15267.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol

ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ADA used to prepare immobilized pH gradients. It can be used to study biological buffers and zwitterionic compounds. It has been used in a study to describe the application of scanning capacitively coupled contactless conductivity detection (SC(4)D) for the determination of pH dependant behaviour of two aminopolycarboxylates immobilized onto the surface of a monolithic capillary column.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26239-55-4
Molecular Formula C6H10N2O5
Molecular Weight (g/mol) 190.155
MDL Number MFCD00008031
InChI Key QZTKDVCDBIDYMD-UHFFFAOYSA-N
Synonym n-2-acetamido iminodiacetic acid, 2,2'-2-amino-2-oxoethyl azanediyl diacetic acid, carbamoylmethylaminodiacetic acid, ada, 2,2'-2-amino-2-oxoethyl imino diacetic acid, n-carbamoylmethyl iminodiacetic acid, glycine, n-2-amino-2-oxoethyl-n-carboxymethyl, n-carbamoylmethyliminodi acetic acid, unii-5c4r3o704e
PubChem CID 117765
ChEBI CHEBI:43960
IUPAC Name 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)O

Specifications

Melting Point ∼218°C (decomposition)
Quantity 25 g
Beilstein 1787181
Merck Index 14,147
Solubility Information Insoluble in water.
Formula Weight 190.16
Percent Purity ≥98%
Chemical Name or Material ADA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.