missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACYP1 Antibody (004), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90125F
This item is not returnable.
View return policy
Description
ACYP1 Monoclonal specifically detects ACYP1 in Human samples. It is validated for ELISA.
Specifications
| ACYP1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 97 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 4 | |
| ELISA | |
| acylphosphatase 1, erythrocyte (common) type, Acylphosphatase, erythrocyte isozyme, Acylphosphatase, organ-common type isozyme, Acylphosphate phosphohydrolase 1, ACYPEacylphosphatase-1, EC 3.6.1.7 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ACYP1 (P07311; Ala 2-Lys 99). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction