missing translation for 'onlineSavingsMsg'
Learn More

Acyclovir, Tocris Bioscience™

Product Code. 18085452
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
18085452 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 18085452

Brand: Tocris Bioscience 2513/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibits viral DNA polymerase; antiherpetic agent

Antiviral agent, active against herpes simplex viruses HSV-1 and HSV-2 (EC50 values are 0.85 and 0.86 μM respectively). Interferes with viral DNA polymerization through competitive inhibition with guanosine triphosphate. Induces apoptosis in cells transfected with HSV-TK (suicidal gene therapy).
TRUSTED_SUSTAINABILITY

Specifications

CAS 59277-89-3
Molecular Formula C8H11N5O3
MDL Number MFCD00057880
Quantity 50 mg
Synonym acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum
InChI Key MKUXAQIIEYXACX-UHFFFAOYSA-N
SMILES NC1=NC2=C(N=CN2COCCO)C(=O)N1
IUPAC Name 2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-1H-purin-6-one
Molecular Weight (g/mol) 225.21
PubChem CID 2022
ChEBI CHEBI:2453
Formula Weight 225.21
Percent Purity >99%
Chemical Name or Material Acyclovir
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.