missing translation for 'onlineSavingsMsg'
Learn More

Acyclovir, MedChemExpress

Product Code. 16477296
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477296 100 mg 100mg
16463347 50 mg 50mg
16461856 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477296 Supplier Medchem Express Supplier No. HY17422
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Acyclovir (Aciclovir) is a potent, orally active antiviral agent. Acyclovir has antiherpetic activity with IC50 values of 0.85 μM and 0.86 μM for HSV-1 and HSV-2, respectively. Acyclovir induces cell cycle perturbation and apoptosis. Acyclovir prevents bacterial infections during induction therapy for acute leukaemia.

Acyclovir (Aciclovir) is a potent, orally active antiviral agent. Acyclovir has antiherpetic activity with IC50 values of 0.85 μM and 0.86 μM for HSV-1 and HSV-2, respectively. Acyclovir induces cell cycle perturbation and apoptosis. Acyclovir prevents bacterial infections during induction therapy for acute leukaemia.

Specifications

Chemical Name or Material Acyclovir
Physical Form Solid
CAS 59277-89-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H11N5O3
For Use With (Application) COVID-19-anti-virus
Synonym Aciclovir Acycloguanosine
Solubility Information DMSO : ≥ 50 mg/mL (222.02 mM)
SMILES O=C1NC(N)=NC2=C1N=CN2COCCO
Molecular Weight (g/mol) 225.2
Formula Weight 225.2
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.