missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acyclovir, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Description
Acyclovir (Aciclovir) is a potent, orally active antiviral agent. Acyclovir has antiherpetic activity with IC50 values of 0.85 μM and 0.86 μM for HSV-1 and HSV-2, respectively. Acyclovir induces cell cycle perturbation and apoptosis. Acyclovir prevents bacterial infections during induction therapy for acute leukaemia.
Specifications
Specifications
| Chemical Name or Material | Acyclovir |
| Physical Form | Solid |
| CAS | 59277-89-3 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C8H11N5O3 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | Aciclovir Acycloguanosine |
| Solubility Information | DMSO : ≥ 50 mg/mL (222.02 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction