missing translation for 'onlineSavingsMsg'
Learn More

Acycloguanosine, 98%

Product Code. 15436229
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436229 5 g 5g
15426229 1 g 1g
15446229 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436229 Supplier Thermo Scientific Alfa Aesar Supplier No. J64144.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Inhibits cytomegalovirus replication; induces apoptosis | CAS: 59277-89-3 | C8H11N5O3 | 225.21 g/mol

Inhibits cytomegalovirus replication; induces apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59277-89-3
Molecular Formula C8H11N5O3
Molecular Weight (g/mol) 225.21
MDL Number MFCD00057880
InChI Key MKUXAQIIEYXACX-UHFFFAOYSA-N
Synonym acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum
PubChem CID 2022
ChEBI CHEBI:2453
IUPAC Name 2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2COCCO)C(=O)N1

Specifications

Melting Point 256°C to 260°C
Color White
Quantity 5 g
Merck Index 14,146
Solubility Information Soluble in 1M HCl at 50mg/ml; Soluble in water at 0;7mg/ml; Also soluble in DMSO
Formula Weight 225.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Acycloguanosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.