missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACY3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16786-1-AP-150UL
This item is not returnable.
View return policy
Description
ACY3 protein function plays an important role in deacetylating mercapturic acids in kidney proximal tubules. Also acts on N-acetyl-aromatic amino acids.Specifications
| ACY3 | |
| Polyclonal | |
| Unconjugated | |
| ACY3 | |
| ACY 3, ACY3, Acylase III, Aminoacylase 3, ASPA2, Aspartoacylase 2, HCBP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 293653, 71670, 91703 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5M876, Q91XE4, Q96HD9 | |
| Acy3 | |
| ACY3 Fusion Protein Ag10220 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction