missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACY3 Monoclonal Antibody (OTI2G4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525389
This item is not returnable.
View return policy
Description
ACY3 Monoclonal Antibody for Western Blot, Flow
ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.
Specifications
| ACY3 | |
| Monoclonal | |
| 0.42 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96HD9 | |
| Acy3 | |
| Full length human recombinant protein of ACY3 produced in 293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2G4 | |
| Unconjugated | |
| Acy3 | |
| 0610006H10Rik; AA3; AAIII; Acy3; ACY-3; Acylase III; aminoacylase 3; Aminoacylase III; Aminoacylase-3; Aspa2; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; aspartoacylase-2; aspartoacylase-3; AW107362; EGK_05921; HCBP1; HCV core-binding protein 1; hepatitis C virus core-binding protein 1; LOW QUALITY PROTEIN: N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); RGD1305394 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 91703 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu