missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACY3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | ACY3 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621788
|
Novus Biologicals
NBP2-48895-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671317
|
Novus Biologicals
NBP2-48895 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACY3 Polyclonal antibody specifically detects ACY3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ACY3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2), 40% Glycerol | |
| 91703 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ACY-3aspartoacylase-2, Acylase III, Aminoacylase-3, aspartoacylase (aminocyclase) 3, EC 3.5.1.-, EC 3.5.1.15, HCBP1ASPA2, Hepatitis C virus core-binding protein 1, MGC9740 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MCSLPVPQEPLRRVAVTGGTHGNEMSGVYLARHWLHAPAELQRASFSAVPVLANPAATSGCRRYVDHDLNRTFTSSFLNSR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title