missing translation for 'onlineSavingsMsg'
Learn More

ACY-957, MedChemExpress

Product Code. 16496637
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496637

Brand: Medchem Express HY104008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ACY-957 is an orally active and selective inhibitor of HDAC1 and HDAC2, with IC50s of 7 nM, 18 nM, and 1300 nM against HDAC1/2/3, respectively, and shows no inhibition on HDAC4/5/6/7/8/9.

ACY-957 is an orally active and selective inhibitor of HDAC1 and HDAC2, with IC50s of 7 nM, 18 nM, and 1300 nM against HDAC1/2/3, respectively, and shows no inhibition on HDAC4/5/6/7/8/9.

Specifications

Chemical Name or Material ACY-957
CAS 1609389-52-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H23N5OS
Quantity 5 mg
Solubility Information DMSO : 83.33 mg/mL (194.00 mM; Need ultrasonic)
SMILES O=C(C1=CC=C2N=C(N3CCNCC3)C=CC2=C1)NC4=CC(C5=CC=CS5)=CC=C4N
Molecular Weight (g/mol) 429.54
Formula Weight 429.54
Percent Purity 98.03%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.