missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ACY-738, 95%

Product Code. 17151126 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
17151126 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 17151126

Brand: Thermo Scientific™ 467911000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material ACY-738
CAS 1375465-91-0
Synonym N-Hydroxy-2-((1-phenylcyclopropyl)amino)pyrimidine-5-carboxamide
Quantity 100 mg
Molecular Formula C14 H14 N4 O2
Formula Weight 270.29
Physical Form Solid
Product Type Reagent
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.