missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACVRL1 Recombinant Human Monoclonal Antibody (6G7)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560718
This item is not returnable.
View return policy
Beskrivning
ACVRL1 Recombinant Monoclonal Antibody for ELISA
Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
Specifikationer
| ACVRL1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P37023 | |
| ACVRL1 | |
| Recombinant Human ACVRL1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2 |
| ELISA | |
| 6G7 | |
| Unconjugated | |
| ACVRL1 | |
| activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 94 | |
| -20°C or -80°C if preferred | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering