Learn More
ACVRL1 Mouse anti-Human, Unconjugated, Clone: 2E11B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66857-1-IG-150UL
Description
The ACVRL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ACVRL1. This antibody recognizes human, mouse antigen. The ACVRL1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| ACVRL1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, Immunohistochemistry | |
| 2E11B10 | |
| Unconjugated | |
| 150 μL | |
| 94 | |
| IgG1 |