missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human ACVRL1 (ALK1), GST Tag Recombinant Protein

Product Code. 15468773
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
1mg
Each
This item is not returnable. View return policy

Product Code. 15468773

Brand: Thermo Scientific™ PV4884

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P37023
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 150mM NaCl, 0.5mM EDTA, 0.02% polysorbate 20, 50% glycerol, 2mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 94
Molecular Weight (g/mol) 68.9 kDa
Name Human ACVRL1 (ALK1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I
Common Name ACVRL1
Gene Symbol ACVRL1
KinaseFamily STKR Kinase Family
Protein Length 139-503
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 139-503
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Catalytic
Form Liquid
Research Category Drug Development
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.