missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACVR2B Recombinant Rabbit Monoclonal Antibody (K01_2L28)

Product Code. 30825826
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825826 50 μL
30828617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825826 Supplier Invitrogen™ Supplier No. MA565447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MTAPWVALAL LWGSLCAGSG RGEAETRECI YYNANWELER TNQSGLERCE GEQDKRLHCY ASWRNSSGTI ELVKKGCWLD DFNCYDRQEC VATEENPQVY Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%, Pig - 98%, Cynomolgus monkey - 99%.

ACVR2B is a type II member of the TGF-beta family of receptor ser/thr kinases and is involved in the activin and myostatin signaling pathway. Activins and inhibins are members of the TGF-beta superfamily due to amino acid homology with respect to the conservation of 7 of the 9 cysteine residues common to all TGF-beta forms. To date, seven type I and five type II activin receptors have been cloned from mammals, including activin receptor IA, activin receptor IIA, activin receptor IB, and activin receptor IIB. In addition, two splice variants of activin receptor IIA and five splice variants of activin receptor IIB have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR2B
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2L28
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ACVR2B
Gene Accession No. P27040, P38445, Q13705
Gene Alias activin A receptor type 2B; activin A receptor type IIB; activin A receptor, type IIB; activin receptor IIB; activin receptor type IIB; Activin receptor type-2B; Activine receptor 2b (transmembrane serine kinase); ActRIIB; ACTR-IIB; ActRIIB type II activin receptor B; ACVR2B; HTX4; MGC116908
Gene Symbols ACVR2B
Host Species Rabbit
Immunogen A synthetic peptide of human Activin Receptor Type IIB/ACVR2B (aa 1-100).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100755103, 11481, 25366, 93
Target Species Human, Mouse, Rat, Hamster
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.