missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACVR1C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12610-1-AP-150UL
This item is not returnable.
View return policy
Description
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I and two type II receptors. ACVR1C, also known as ALK7, is a type I activin receptor and plays a role in cell differentiation, growth arrest and apoptosis. ACVR1C can mediate signaling by ligans such as Nodal, GDF-1/3, activin B and activin AB, all of which can also signal through the ubiquitous activin type I receptor ACVR1B (also known as ALK4). ACVR1C is a novel marker specifically expressed during the late phase of adipocyte differentiation. ACVR1C is dispensable for mouse embryogenesis, which suggests alternative functions for this receptor in postnatal development and tissue homeostasis. ACVR1C plays an important role in regulating the functional plasticity of pancreatic islets, negatively affecting beta-cell function by mediating the effects of activin B on Ca2+ signaling.Specifications
| ACVR1C | |
| Polyclonal | |
| Unconjugated | |
| ACVR1C | |
| activin A receptor, type IC, Activin receptor like kinase 7, Activin receptor type 1C, Activin receptor type IC, ACTR IC, ACVR1C, ACVRLK7, ALK 7, ALK7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 130399, 245921, 269275 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P70539, Q8K348, Q8NER5 | |
| ACVR1C | |
| ACVR1C Fusion Protein Ag3162 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction