Learn More
Invitrogen™ ACVR1C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533317
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine, Bat, Pig (100%); Marmoset, Elephant, Panda, Horse, Rabbit (94%); Opossum, Turkey, Chicken, Platypus (88%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment with Proteinase K.
ACVR1C, a serine/threonine protein kinase, is a type I receptor for the TGFB family of signaling molecules. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors. ACVR1C is a receptor for activin AB, activin B and NODAL. This protein plays a role in cell differentiation, growth arrest and apoptosis.
Specifications
| ACVR1C | |
| Polyclonal | |
| Unconjugated | |
| ACVR1C | |
| activin A receptor type 1C; activin A receptor type IC; activin A receptor, type IC; activin receptor type IC; Activin receptor type-1C; activin receptor-like kinase 7; ACTRI; ACTR-IC; ACVR1A; acvr1c; acvr1c {ECO:0000312; ACVRLK2; ACVRLK7; ALK2; Alk7; ALK-7; C230097P10; EMBL:AAC52803.1}; FOP; habrec1; SKR1; TGF-beta type 1 receptor; TSRI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100156756, 130399, 536380 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8NER5 | |
| ACVR1C | |
| Synthetic 16 amino acid peptide from internal region of human ACVR1C. | |
| 50 μg | |
| Primary | |
| Bat, Bovine, Human, Pig | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.