missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACVR1B Recombinant Rabbit Monoclonal Antibody (K01_1M35)

Product Code. 30828932
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828932 50 μL
30829914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828932 Supplier Invitrogen™ Supplier No. MA564643

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ACVR1B Recombinant Monoclonal Antibody for Western Blot

ACVR1B is a serine/threonine kinase involved in signal transduction and cellular communication. Several truncated ACVR1B isoforms have been reported to be exclusively expressed in human pituitary tumors. Mutations of ACVR1B have been observed in pancreatic adenocarcinomas. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. ACVR1B encodes activin A type IB receptor, composed of 11 exons. Alternative splicing and alternative polyadenylation result in 3 fully described transcript variants. The mRNA expression of variants 1, 2, and 3 is confirmed, and a potential fourth variant contains an alternative exon 8 and lacks exons 9 through 11, but its mRNA expression has not been confirmed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR1B
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1M35
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ACVR1B
Gene Accession No. P36896, P80202, Q61271
Gene Alias 6820432J04; activin A receptor type 1B; activin A receptor type IB; activin A receptor, type 1B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; activin receptor type IB; Activin receptor type-1B; activin receptor-like kinase 4; ACTRI; ActRIB; ActR-IB; ACVR1A; Acvr1b; ACVRLK2; ACVRLK4; ALK2; Alk4; ALK-4; FOP; Serine/threonine-protein kinase receptor R2; SKR1; SKR2; TSRI
Gene Symbols ACVR1B
Host Species Rabbit
Immunogen A synthetic peptide of human Activin A Receptor Type IB.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11479, 29381, 91
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.