missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACVR1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596328
This item is not returnable.
View return policy
Description
Immunogen sequence: SGPRGVQALL CACTSCLQAN YTCETDGACM VSIFNLDGME HHVRTCIPKV ELVPAGKPFY CLSSEDLRNT HCCYTDYCNR IDLRVPSGHL KEPEHPSMWG PVE; Positive Samples: SW480, Mouse kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein.
ACVR1B is a serine/threonine kinase involved in signal transduction and cellular communication. Several truncated ACVR1B isoforms have been reported to be exclusively expressed in human pituitary tumors. Mutations of ACVR1B have been observed in pancreatic adenocarcinomas. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. ACVR1B encodes activin A type IB receptor, composed of 11 exons. Alternative splicing and alternative polyadenylation result in 3 fully described transcript variants. The mRNA expression of variants 1, 2, and 3 is confirmed, and a potential fourth variant contains an alternative exon 8 and lacks exons 9 through 11, but its mRNA expression has not been confirmed.
Specifications
| ACVR1B | |
| Polyclonal | |
| Unconjugated | |
| ACVR1B | |
| 6820432J04; activin A receptor type 1B; activin A receptor type IB; activin A receptor, type 1B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; activin receptor type IB; Activin receptor type-1B; activin receptor-like kinase 4; ACTRI; ActRIB; ActR-IB; ACVR1A; Acvr1b; ACVRLK2; ACVRLK4; ALK2; Alk4; ALK-4; FOP; Serine/threonine-protein kinase receptor R2; SKR1; SKR2; TSRI | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11479, 91 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.56 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36896, Q61271 | |
| ACVR1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-126 of human ACVR1B (NP_0042931). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction