missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACVR1B Monoclonal Antibody (179CT11.2.1)

ACVR1B Monoclonal Antibody (179CT11.2.1)
Brand: Thermo Scientific MA537487
This item is not returnable.
View return policy
Description
ACVR1B Monoclonal Antibody for Western Blot
Specifications
| ACVR1B | |
| Monoclonal | |
| Unconjugated | |
| ACVR1B | |
| 6820432J04; activin A receptor type 1B; activin A receptor type IB; activin A receptor, type 1B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; activin receptor type IB; Activin receptor type-1B; activin receptor-like kinase 4; ACTRI; ActRIB; ActR-IB; ACVR1A; Acvr1b; ACVRLK2; ACVRLK4; ALK2; Alk4; ALK-4; FOP; Serine/threonine-protein kinase receptor R2; SKR1; SKR2; TSRI | |
| Mouse | |
| Protein G | |
| RUO | |
| 11479 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 179CT11.2.1 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q61271 | |
| ACVR1B | |
| ACVR1B recombinant protein. | |
| 400 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction