Learn More
Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes
Product Code. 10263152
Change view
Click to view available options
Volume (Metric):
1 μL
5 μL
10 μL
15 μL
20 μL
25 μL
40 μL
50 μL
60 μL
70 μL
75 μL
80 μL
90 μL
100 μL
120 μL
150 μL
200 μL
250 μL
300 μL
400 μL
500 μL
600 μL
700 μL
750 μL
800 μL
900 μL
1000 μL
Unit Size:
Each
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10263152 50 μL Each
10355122 100 μL Each
10161582 200 μL Each
10724613 250 μL Each
10733463 1000 μL Each
10754623 1 μL Each
10467072 40 μL Each
10632673 60 μL Each
10375022 70 μL Each
10551523 75 μL Each
10121442 80 μL Each
10345122 90 μL Each
10591903 120 μL Each
10632863 150 μL Each
10375262 300 μL Each
10485152 400 μL Each
10365312 500 μL Each
10089651 600 μL Each
10099651 700 μL Each
10191722 750 μL Each
10315412 800 μL Each
10099721 900 μL Each
10162062 5 μL Each
10611333 10 μL Each
10764623 15 μL Each
10541333 20 μL Each
10447652 25 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Product Code. 10263152 Supplier Socorex™ Supplier No. 815.0050
Only null left
Add to basket
Add to basket
Slim and precise micropipettes set at a fixed volume for easy dose repetition. Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes are easy to handle and have a soft plunger stroke.
  • Air-displacement fixed-volume pipette.
    • Exceptional, user-friendly ergonomics.
    • Justip™ adjustable tip ejector fitting most tips.
    • Shock, UV-light and autoclaving resistance.
    • Ultra-soft plunger stroke.
    • Available in a variety of volume models.
Specifications
Accuracy <±0.7%
Certifications/Compliance ISO 8655, GMP, GLP, NCCLS, CE IVD Directive 98/79 EEC (in vitro diagnostic medical devices).
Autoclavable Autoclavable
Color Yellow
Model 815
Type Micropipette
Volume (Metric) 50 μL
Precision <0.5%
Quantity 1 Each
Warranty 36 Months