missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura™ Electro 936 Macropipette
Ergonomically friendly, this electronic single-channel macropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 936 Macropipette charges quickly and has a long working autonomy.
£570.00
Specifications
Battery Type | 4.8V NiMH |
---|---|
Certifications/Compliance | EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) |
Model | Acura™ Electro™ 936 |
Number of Channels | 1 |
Product Line | Acura™ |
Product Code | Brand | Accuracy | Precision | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Precision | Volume (Metric) | Price | Quantity & Availability | |||
10050992
|
Socorex™
936.10 |
<±1.0% - <±0.5% | <0.5% - <0.15% | 0.5 to 10 mL |
£570.00
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11889223
|
Socorex™
936.05 |
<±1.2% - <±0.5% | <0.6% - <0.15% | 0.25 to 5 mL |
£570.00
Each |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Interchangeable nozzle protection filter.
- Adapters for Pasteur pipette and straw tips.
- Outstanding accuracy and precision.
Specifications
4.8V NiMH | |
Acura™ Electro™ 936 | |
Acura™ | |
Right/Left LCD | |
0.05mL | |
<1.5Hours |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
1 | |
Electronic Macropipette | |
QC certificate, Qualitips™ pipette tip samples and operating instructions. | |
1 Each | |
24 Months |