missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura™ Electro 926 Electronic Single-Channel Micropipette Initial Kit
Ergonomic electronic single-channel micropipette initial package. Acura™ Electro 926 Electronic Single-Channel Micropipette Initial Kit includes pipette, charging stand, fast charger, AC adapter and cord, additional battery pack, and pipette tip samples.
£795.00
Specifications
Battery Type | 4.8 V NiMH |
---|---|
Certifications/Compliance | EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) |
Model | 926 |
Number of Channels | 1 |
Product Line | Acura |
Product Code | Brand | Volume (Metric) | Increments | Accuracy | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Increments | Accuracy | Price | Quantity & Availability | |||
11819213
|
Socorex™
926.0002E |
0.1 to 2.0 μL | 0.01 μL | <±2.5% to <±0.9% |
£795.00
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
11899213
|
Socorex™
926.0050E |
2.5 to 50 μL | 0.25μL | <±1.0% to <±0.5% |
£795.00
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
11859223
|
Socorex™
926.1000E |
50 to 1000 μL | 5 μL | <±0.8% to <±0.4% |
£795.00
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
- Includes pipetting cycle counter.
- Optional volumetric modules.
Specifications
4.8 V NiMH | |
926 | |
Acura | |
Right/Left LCD | |
1 Ea. | |
24 Months |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
1 | |
Starter Kit, Micro | |
Pipette charging stand, fast charger with AC adapter and cord, additional battery pack, QC certificate, Qualitips™ pipette tip samples and operating instructions. | |
<1.5 hr. |