missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura™ Electro 926 Electronic Single-Channel Micropipette Initial Kit
Click to view available options
Volume (Metric):
0.1 to 2.0 μL
2.5 to 50 μL
50 to 1000 μL
Unit Size:
Each
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
- Includes pipetting cycle counter.
- Optional volumetric modules.
Specifications
Specifications
| Accuracy | <±0.8% to <±0.4% |
| Battery Type | 4.8 V NiMH |
| Certifications/Compliance | EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) |
| Model | 926 |
| Number of Channels | 1 |
| Product Line | Acura |
| Type | Starter Kit, Micro |
| Volume (Metric) | 50 to 1000 μL |
| Display | Right/Left LCD |
| Includes | Pipette charging stand, fast charger with AC adapter and cord, additional battery pack, QC certificate, Qualitips™ pipette tip samples and operating instructions. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction