missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTRT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550301
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ACTRT1 protein.
This gene encodes a protein related to the cytoskeletal protein beta-actin. This protein is a major component of the calyx in the perinuclear theca of mammalian sperm heads, and it therefore likely functions in spermatid formation. This gene is intronless and is similar to a related gene located on chromosome 1. A related pseudogene has also been identified approximately 75 kb downstream of this gene on chromosome X.
Specifications
| ACTRT1 | |
| Polyclonal | |
| Unconjugated | |
| ACTRT1 | |
| actin related protein T1; actin-related protein T1; ACTRT1; AIP1; ARIP1; ARPT1; ARP-T1; HSD27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 139741 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8TDG2 | |
| ACTRT1 | |
| Fusion protein corresponding to residues near the C terminal of human Actin-related protein T1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction