missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTR8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113384
This item is not returnable.
View return policy
Description
ACTR8 Polyclonal Antibody for Western Blot, ELISA
ACTR8 plays an important role in the functional organization of mitotic chromosomes.
Specifications
| ACTR8 | |
| Polyclonal | |
| Unconjugated | |
| ACTR8 | |
| 5730542K05Rik; actin related protein 8; actin-related protein 8; actin-related protein 8 homolog; Actr8; Arp8; ARP8 actin related protein 8 homolog; ARP8 actin-related protein 8; ARP8 actin-related protein 8 homolog; ARP8 actin-related protein 8 homolog (yeast); hArp8; INO80 complex subunit N; INO80N | |
| Rabbit | |
| Protein G | |
| RUO | |
| 93973 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H981 | |
| ACTR8 | |
| Recombinant Human Actin-related protein 8 protein (35-175AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction