missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558454
This item is not returnable.
View return policy
Description
Immunogen sequence: NAGALSAHRY FRDNPSELCC IIVDSGYSFT HIVPYCRSKK KKEAIIRINV GGKLLTNHLK EIISYRQLHV MDETHVINQV KEDVCYV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The function of this protein remains unknown.
Specifications
| ACTR6 | |
| Polyclonal | |
| Unconjugated | |
| ACTR6 | |
| 2010200J04Rik; actin related protein 6; actin-related protein 6; actin-related protein 6-like protein; Actr6; Arp6; ARP6 actin related protein 6 homolog; ARP6 actin-related protein 6; ARP6 actin-related protein 6 homolog; ARP6 actin-related protein 6 homolog (yeast); ArpX; AU044443; CDA12; FLJ13433; h hARPX; hArp6; hARPX; HSPC281; LOW QUALITY PROTEIN: actin-related protein 6; mArp6; MSTP136 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9GZN1 | |
| ACTR6 | |
| Recombinant protein corresponding to Human ACTR6. Recombinant protein control fragment (Product #RP-97638). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction