missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTR1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107052
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex.
Specifications
| ACTR1B | |
| Polyclonal | |
| Unconjugated | |
| Actr1b | |
| 2310066K23Rik; AA960180; Actin-related protein 1B; Actr1b; AI851923; ARP1; ARP1 actin-related protein 1 homolog B, centractin beta; ARP1 actin-related protein 1 homolog B, centractin beta (yeast); ARP1 actin-related protein 1B, centractin beta; Arp1b; Beta-centractin; centractin beta; CTRN2; PC3; yeast homolog B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10120 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P42025 | |
| Actr1b | |
| Synthetic peptide directed towards the C-terminal region of human ACTR1B (aa 327-376). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction