missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTR1B Monoclonal Antibody (4E10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520934
This item is not returnable.
View return policy
Description
Peptide Sequence: SRYLRLLLRK EGVDFHTSAE FEVVRTIKER ACYLSINPQK DEALETEKVQ YTLPDGSTLD VGPARFRAPE LLFQPDLVGD ESEGLHEVVA FAIHK*.
This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex.
Specifications
| ACTR1B | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P42025 | |
| Actr1b | |
| ACTR1B (NP_005726, 191 a.a. ~ 286 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry | |
| 4E10 | |
| Unconjugated | |
| Actr1b | |
| 2310066K23Rik; AA960180; Actin-related protein 1B; Actr1b; AI851923; ARP1; ARP1 actin-related protein 1 homolog B, centractin beta; ARP1 actin-related protein 1 homolog B, centractin beta (yeast); ARP1 actin-related protein 1B, centractin beta; Arp1b; Beta-centractin; centractin beta; CTRN2; PC3; yeast homolog B | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10120 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction