missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15275
This item is not returnable.
View return policy
Description
ACTR1B Polyclonal specifically detects ACTR1B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ACTR1B | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| Actin-related protein 1B, ARP1 (actin-related protein 1, yeast) homolog B (centractin beta), ARP1 actin-related protein 1 homolog B, centractin beta (yeast), ARP1BPC3, centractin beta, CTRN2beta-centractin | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Expected to cross react based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.69mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P42025 | |
| ACTR1B | |
| Recombinant protein encompassing a sequence within the center region of human ACTR1B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10120 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction