missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTR1A Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518273
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes a 42.6 kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 8-13 copies per dynactin molecule, and is the most abundant molecule in the dynactin complex. It is an actin-related protein, and is approximately 60% identical at the amino acid level to conventional actin.
Specifications
| ACTR1A | |
| Polyclonal | |
| Unconjugated | |
| ACTR1A | |
| actin related protein 1A; actin-related protein 1 alpha; actin-related protein 1 homolog A; actin-RPV; ACTR1A; actr1a {ECO:0000250; alpha-Arp1; alpha-centractin; ARP1; ARP1 actin related protein 1 homolog A; ARP1 actin-related protein 1 homolog A, centractin alpha; ARP1 actin-related protein 1 homolog A, centractin alpha (yeast); ARP1 actin-related protein 1A, centractin alpha; Arp1 centractin alpha; Arp1 contractin alpha; Arp1A; centractin; Centrosome-associated actin homolog; CTRN1; FLJ52695; FLJ52800; FLJ55002; UniProtKB:P61163}; yeast homolog A | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10121, 294010, 403791, 54130 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P61162, P61163, P61164, P85515 | |
| ACTR1A | |
| Synthetic peptide sequence (YEEDGARSIHRKT) corresponding to the C-terminus amino acids of ACTR1A (aa 363-376). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction