missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | ACTR1A |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226473
|
Novus Biologicals
NBP3-38278-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231222
|
Novus Biologicals
NBP3-38278-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACTR1A Polyclonal antibody specifically detects ACTR1A in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| ACTR1A | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Actin-RPV, alpha-centractin, ARP1 (actin-related protein 1, yeast) homolog A (centractin alpha), ARP1 actin-related protein 1 homolog A, centractin alpha (yeast), ARP1actin-RPV, centractin, Centrosome-associated actin homolog, CTRN1, FLJ52695, FLJ52800, FLJ55002 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 187-376 of human ACTR1A (NP_005727.1).,, Sequence:, GRDVSRFLRLYLRKEGYDFHSSSEFEIVKAIKERACYLSINPQKDETLETEKAQYYLPDGSTIEIGPSRFRAPELLFRPDLIGEESEGIHEVLVFAIQKSDMDLRRTLFSNIVLSGGSTLFKGFGDRLLSEVKKLAPKDVKIRISAPQERLYSTWIGGSILASLDTFKKMWVSKKEYEEDGARSIHRKTF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10121 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title