missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTR10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92268-0.1ml
This item is not returnable.
View return policy
Description
ACTR10 Polyclonal antibody specifically detects ACTR10 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ACTR10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| actin-related protein 10 homolog (S. cerevisiae), Actin-related protein 11, ACTR11actin-related protein 10, Arp11, HARP11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 218-417 of human ACTR10 (NP_060947.1). SDLKRGLKIQAAKFNIDGNNERPSPPPNVDYPLDGEKILHILGSIRDSVVEILFEQDNEEQSVATLILDSLIQCPIDTRKQLAENLVVIGGTSMLPGFLHRLLAEIRYLVEKPKYKKALGTKTFRIHTPPAKANCVAWLGGAIFGALQDILGSRSVSKEYYNQTGRIPDWCSLNNPPLEMMFDVGKTQPPLMKRAFSTEK | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 55860 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction