missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTL7B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£328.00 - £470.00
Specifications
| Antigen | ACTL7B |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18405491
|
Novus Biologicals
NBP1-86973-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18294237
|
Novus Biologicals
NBP1-86973 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACTL7B Polyclonal specifically detects ACTL7B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACTL7B | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 10880 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MATRNSPMPLGTAQGDPGEAGTRPGPDASLRDTGAATQLKMKPRKVHKIKAVIIDLGSQYCKCGYAGEPRPTYFISSTVGKRCPEAADAGDTRKWTL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| Human | |
| actin-like 7B, actin-like 7-beta, actin-like protein 7B, actin-like-7-beta, Tact1 | |
| ACTL7B | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title