missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTL6B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | ACTL6B |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:500-1:1000, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18361777
|
Novus Biologicals
NBP3-18635-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18344632
|
Bio-Techne
NBP3-18635-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACTL6B Polyclonal antibody specifically detects ACTL6B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ACTL6B | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Extracellular Matrix | |
| Whole antiserum | |
| 53 kDa BRG1-associated factor B, actin-like 6, actin-like 6B, Actin-related protein Baf53b, ACTL6, ArpNalpha, BAF53Bactin-like protein 6B, BRG1-associated factor 53B, hArpN alpha | |
| Polyclonal antibody raised in rabbit against human ACTL6B (actin-like 6B), using a KLH-conjugated synthetic peptide containing a sequence from the central part of the protein. | |
| Whole Antiserum |
| Western Blot 1:1000, ELISA 1:500-1:1000, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| O94805 | |
| 51412 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title