missing translation for 'onlineSavingsMsg'
Learn More

Activin RIIA Rabbit anti-Human, Mouse, Rat, Clone: 1K1J10, Novus Biologicals™

Product Code. 18348013
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348013 20 μg 20µL
18383284 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18348013 Supplier Bio-Techne Supplier No. NBP31659820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Activin RIIA Monoclonal antibody specifically detects Activin RIIA in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Activin RIIA
Applications Western Blot
Classification Monoclonal
Clone 1K1J10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ACTRII, ACTRIIA, ACVR2ACTR-IIA, EC 2.7.11, EC 2.7.11.30
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Activin RIIA (P27037).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 92
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.