missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Activin RIIA Antibody (Ab-14E1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28281-50ug
This item is not returnable.
View return policy
Description
Activin RIIA Monoclonal antibody specifically detects Activin RIIA in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Activin RIIA | |
| Monoclonal | |
| Unconjugated | |
| P27037 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Ab-14E1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ACTRII, ACTRIIA, ACVR2ACTR-IIA, EC 2.7.11, EC 2.7.11.30 | |
| ACVR2A | |
| 50 μg | |
| Cancer | |
| 92 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction