missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin RIB/ALK-4 Rabbit anti-Human, Mouse, Rat, Clone: 3U6I8, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16106-100UL
This item is not returnable.
View return policy
Description
Activin RIB/ALK-4 Monoclonal antibody specifically detects Activin RIB/ALK-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Activin RIB/ALK-4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3U6I8 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ACTRIB, ACVRLK4ACTR-IB, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Activin RIB/ALK-4 (P36896). | |
| 100 μg | |
| Protein Kinase | |
| 91 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction