missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin RIB/ALK-4 Antibody (JE38-48), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-31998
This item is not returnable.
View return policy
Description
Activin RIB/ALK-4 Monoclonal antibody specifically detects Activin RIB/ALK-4 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Activin RIB/ALK-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250, Immunohistochemistry-Paraffin 1:5000 | |
| activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ACTRIB, ACVRLK4ACTR-IB, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B | |
| Recombinant protein within human Activin RIB/ALK-4 aa 24-126. (Uniprot: P36896) | |
| 100 μL | |
| Protein Kinase | |
| 91 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| JE38-48 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction